5-(Chloromethyl)-3-methyl-1H-pyrazole hydrochloride

Toimittaja: Apollo Scientific

APOSOR310728-1GEA 525 EUR
APOSOR310728-1G
5-(Chloromethyl)-3-methyl-1H-pyrazole hydrochloride
5-(Chloromethyl)-3-methyl-1H-pyrazole hydrochloride
Kaava: C₅H₇ClN₂·ClH
MW: 167,036 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD28023517
CAS-numero: 278798-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR