2',3',4'-Trihydroksiasetofenoni

APOSBIG1001-100GEA 0 EUR
APOSBIG1001-100G APOSBIG1001-25G
2',3',4'-Trihydroksiasetofenoni
2',3',4'-Trihydroksiasetofenoni
Kaava: C₈H₈O₄
MW: 168,15 g/mol
Sulamispiste: 169…172 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002193
CAS-numero: 528-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR