Biotin-PEG2-NHS

APOSBIPG1228-250MGEA 0 EUR
APOSBIPG1228-250MG
Biotin-PEG2-NHS
Biotin-PEG2-NHS
Kaava: C₂₁H₃₂N₄O₈S
MW: 500,57 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00467154
CAS-numero: 596820-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR