2-(Benzylamino)-4-thiazolecarboxylic acid

Varoitus

APOSOR12022-5GEA 0 EUR
APOSOR12022-5G APOSOR12022-1G
2-(Benzylamino)-4-thiazolecarboxylic acid
2-(Benzylamino)-4-thiazolecarboxylic acid
Kaava: C₁₁H₁₀N₂O₂S
Storage Temperature: Jääkaappi
MDL numero: MFCD06655105
CAS-numero: 165682-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR