3-Amino-3-(3-methoxyphenyl)propionic acid

Varoitus

APOSOR13354-1GEA 26.4 EUR
APOSOR13354-1G
3-Amino-3-(3-methoxyphenyl)propionic acid
3-Amino-3-(3-methoxyphenyl)propionic acid
Kaava: C₁₀H₁₃NO₃
MW: 195,22
Storage Temperature: Jääkaappi
MDL numero: MFCD00462569
CAS-numero: 68208-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR