2,4-Dichlorophenacyl thiocyanate

APOSOR14050-1GEA 0 EUR
APOSOR14050-1G APOSOR14050-5G APOSOR14050-10G
2,4-Dichlorophenacyl thiocyanate
2,4-Dichlorophenacyl thiocyanate
Kaava: C₉H₅Cl₂NOS
Sulamispiste: 100…102 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03453108
CAS-numero: 125488-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR