2-(4-Methoxyphenyl)-1,3-thiazolane

APOSOR14062-1GEA 0 EUR
APOSOR14062-1G APOSOR14062-5G APOSOR14062-10G
2-(4-Methoxyphenyl)-1,3-thiazolane
2-(4-Methoxyphenyl)-1,3-thiazolane
Kaava: C₁₀H₁₃NOS
Sulamispiste: 100…102 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00222821
CAS-numero: 31404-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR