5-(Tributylstannyl)pyrimidine

APOSOR15531-1GEA 185 EUR
APOSOR15531-1G APOSOR15531-5G
5-(Tributylstannyl)pyrimidine
5-(Tributylstannyl)pyrimidine
Kaava: C₁₆H₃₀N₂Sn
MW: 369,13 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01319056
CAS-numero: 144173-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR