3-Methyl-2-(tributylstannyl)pyridine 97%

Vaara

APOSOR15616-1GEA 277 EUR
APOSOR15616-1G APOSOR15616-250MG APOSOR15616-5G
3-Methyl-2-(tributylstannyl)pyridine 97%
3-Methyl-2-(tributylstannyl)pyridine
Kaava: C₁₈H₃₃NSn
MW: 382,16 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07787404
CAS-numero: 259807-97-1
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR