1,2,5-Trimethylpyrrole-3-carboxylic acid

Varoitus

APOSOR23110-250MGEA 23.8 EUR
APOSOR23110-250MG APOSOR23110-1G
1,2,5-Trimethylpyrrole-3-carboxylic acid
1,2,5-Trimethylpyrrole-3-carboxylic acid
Kaava: C₈H₁₁NO₂
Sulamispiste: 150 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00203866
CAS-numero: 175276-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR