Methyl-4-formyl-1,2,5-trimethylpyrrole-3-carboxylate

Varoitus

APOSOR23358-1GEA 0 EUR
APOSOR23358-1G APOSOR23358-250MG
Methyl-4-formyl-1,2,5-trimethylpyrrole-3-carboxylate
Methyl-4-formyl-1,2,5-trimethylpyrrole-3-carboxylate
Kaava: C₁₀H₁₃NO₃
Sulamispiste: 124…127 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00203864
CAS-numero: 175276-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR