1,4-Diiodo-2,5-dimethoxybenzene

Varoitus

APOSOR2448-1GEA 26.4 EUR
APOSOR2448-1G APOSOR2448-10G APOSOR2448-5G
1,4-Diiodo-2,5-dimethoxybenzene
1,4-Diiodo-2,5-dimethoxybenzene
Kaava: C₈H₈I₂O₂
Sulamispiste: 171…173 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00461369
CAS-numero: 51560-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR