2-Amino-9H-pyrido[2,3-b]indole

Varoitus

APOSOR2900T-10MGEA 0 EUR
APOSOR2900T-10MG
2-Amino-9H-pyrido[2,3-b]indole
2-Amino-9H-pyrido[2,3-b]indole

A pyrolysate that exhibits mutagenic activity toward Salmonella typhimurium TA98 & TA100.

Kaava: C₁₁H₉N₃
MW: 183,21 g/mol
Sulamispiste: 194…196 °C
Storage Temperature: Pakastin
MDL numero: MFCD00210750
CAS-numero: 26148-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR