3-Chloro-4-phenyl-1,2,5-thiadiazole

APOSOR322474-250MGEA 0 EUR
APOSOR322474-250MG
3-Chloro-4-phenyl-1,2,5-thiadiazole
3-Chloro-4-phenyl-1,2,5-thiadiazole
Kaava: C₈H₅ClN₂S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01935960
CAS-numero: 5728-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR