4-Methoxy-2-methylbenzonitrile 97%

APOSOR43646-25GEA 0 EUR
APOSOR43646-25G APOSOR43646-10G
4-Methoxy-2-methylbenzonitrile 97%
4-Methoxy-2-methylbenzonitrile
Kaava: C₉H₉NO
MW: 147,1700 g/mol
Sulamispiste: 177…181 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03425692
CAS-numero: 21883-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR