Cyclohexyl propionate 95%

Toimittaja: Apollo Scientific

APOSOR918728-25GEA 148 EUR
APOSOR918728-25G
Cyclohexyl propionate 95%
Cyclohexyl propionate
Kaava: C₉H₁₆O₂
MW: 156,22 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00046352
CAS-numero: 6222-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR