Methyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%
Toimittaja: Apollo Scientific
APOSOR965437-1GEA
171
EUR
APOSOR965437-1G
Methyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 95%
Methyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Kaava:
C₁₀H₁₀N₂O₂ MW: 190,2000 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD15203666 CAS-numero: 943112-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...