Pentafluorophenyl phenylmethanesulphonate

Toimittaja: Apollo Scientific
Varoitus

APOSPC10318-1GEA 134 EUR
APOSPC10318-1G
Pentafluorophenyl phenylmethanesulphonate
Pentafluorophenyl phenylmethanesulphonate
Kaava: C₁₃H₇F₅O₃S
Sulamispiste: 91…93 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05975135
CAS-numero: 864450-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR