SuperCos I vector
Toimittaja: Agilent Technologies
AGLS251301EA 673 EUR
AGLS251301
SuperCos I vector
SuperCos 1 is a novel, 7,9 kb cosmid vector that contains bacteriophage promoter sequences flanking a unique cloning site.
  • Cloning capacity of 30 to 42 kb
  • For generating cosmid libraries
  • Ideal for high-resolution restriction mapping and rapid chromosomal walking
This structure allows rapid synthesis of 'walking' probes specific for the extreme ends of insert DNA.
The SuperCos 1 vector is also engineered to contain genes for the amplification and expression of cosmid clones in eukaryotic cells.
Varoitus: For research use only. Not for use in diagnostic procedures.