DMEM, high glucose, stable glutamine, pyruvate
Toimittaja: Apollo Scientific
APOSBINC302-1PKEA 0 EUR
APOSBINC302-1PK APOSBINC302-10PK
DMEM, high glucose, stable glutamine, pyruvate
Soluviljelyyn liuokset
With glucose (4500 mg/L), stable L-glutamine (870 mg/L), sodium bicarbonate (3700 mg/L) + phenol red.