Achieve consistent high specificity during qRT-PCR detection of miRNA in real time.
- Achieve accurate real-time PCR quantification for miRNA
- A high-specificity reagent system that delivers consistent, sensitive and reproducible results
- The kit elongates miRNAs in a polyadenylation reaction and then reverse transcribes the polyadenylated RNA into QPCR-ready cDNA
- The reagents included enable the synthesis of first-strand cDNA from the tailed miRNAs
- The kit master mix contains all of the necessary enzymes, dyes (including EvaGreen® Dye) and reagents to carry out accurate qRT-PCR detection of microRNA
Our miRNA qRT-PCR detection system enables accurate and real-time PCR quantification for miRNA. The kit is a high-specificity reagent system that delivers sensitive and reproducible results. The kit elongates miRNAs in a polyadenylation reaction and then reverse transcribes the polyadenylated RNA into QPCR-ready cDNA.
The reagents provided enable the synthesis of first-strand cDNA from the tailed miRNAs. In addition, the master mix contains all of the necessary enzymes, dyes (including EvaGreen Dye), and reagents to carry out accurate qRT-PCR detection of microRNA.
Varoitus: For research use only. Not for use in diagnostic procedures.