Taq DNA polymerase, SureStart
Toimittaja: Agilent Technologies
SureStart
AGLS600284EA 1123 EUR
AGLS600284
Taq DNA polymerase, SureStart
Nukleiinihapporeagenssit End-point PCR Enzymes and Kits
Easily integrates into PCR protocols optimized with Taq DNA polymerase, with little or no modification.
  • Eliminates amplification of background artifacts
  • Minimises smearing in long PCR
  • Hot start increases specificity and sensitivity
  • Reliable room-temperature setup
SureStart Taq DNA polymerase is a hot start Taq DNA polymerase. SureStart Taq can be incorporated into PCR protocols previously optimised with Taq DNA polymerase, with little or no modification of cycling parameters or reaction conditions.
Varoitus: For research use only. Not for use in diagnostic procedures.