T3 and T7 polymerases
Toimittaja: Agilent Technologies
AGLS600124EA 519 EUR
AGLS600124 AGLS600123
T3 and T7 polymerases
Nukleiinihapporeagenssit Nucleic Acid Modification and Labelling
DNA-dependent RNA polymerases with high sequence specificity for T3 or T7 promoters.
  • Probes for nucleic acid hybridisations
  • Templates for in vitro translations
  • Substrates for RNA processing studies
  • Exon and intron mapping of genomic DNA
Bacteriophage T3 and T7 RNA polymerases are DNA-dependent RNA polymerases with high sequence specificity for T3 or T7 promoters. Both T3 and T7 RNA polymerases synthesise RNA 5' to 3' and incorporate 35S, 32P, and 33P ribonucleotides. These enzymes are isolated from an overproducing recombinant E. coli clone.
Varoitus: For research use only. Not for use in diagnostic procedures.