RAP top-up kit
Toimittaja: Oxford Nanopore Technologies
OXNTEXP-RAP001EA 420 EUR
OXNTEXP-RAP001
RAP top-up kit
Nukleiinihapporeagenssit Next Generation Sequencing -reagenssit
Rapid Sequencing Adapters (RAP).
The RAP Top-Up Kit contains the Rapid Adapter (RAP) Sequencing Buffer (SQB), Sequencing Tether (SQT), and Loading Beads (LB), sufficient for six sequencing libraries. This kit is compatible for use with the following kits, 16S Barcoding Kit (OXNTSQK-RAB204), 16S Barcoding Kit 1-24 (OXNTSQK-16S024), Rapid Barcoding Kit (OXNTSQK-RBK00), Rapid PCR Barcoding Kit (OXNTSQK-RPB004), PCR Barcoding Kit (OXNTSQK-PBK004) and Flow Cell Wash Kit (OXNTEXP-WSH004).
This product includes a flow cell priming kit.
Tilaustiedot: This product is anticipated to be discontinued in September 2023.
Toimitustiedot: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Varoitus: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.