Safety goggle, Cosmos™
Toimittaja: JSP
Cosmos™
JSPA2COS23CEA 18.8 EUR
JSPA2COS23C
Safety goggle, Cosmos™
Suojalasit, naamiomalliset
Safety goggle offers a wide field of vision and can be worn over prescription frames.
  • Soft browguard
  • Adjustable strap
  • Can be worn over prescription frames
  • Indirect ventilation
The Cosmos™ Safety Goggle, with soft browguard and adjustable straps, has a wide field of vision and can be worn over prescription frames. Improved ergonomics, thanks to the bi-material injection, ensures optimum comfort and resistance to wear.
Sertifikaatit: Meets EN 166:2001.