Two-Way PVC Ball Valves, Female NPT
Toimittaja: Avantor Fluid Handling
MFLX07387-62EA 50.1 EUR
MFLX07387-62 MFLX07387-42 MFLX07387-22
Two-Way PVC Ball Valves, Female NPT
Venttiilit Palloventtiilit
Create a secure pipe threaded connection.
  • Constructed of durable PVC
Control flow with the 90° rotating handle. Valves have seals made of Viton® fluoroelastomer and seats made of PTFE. Maximum pressure is 150 psi at 32 °F (10.3 bar at 0 °C); maximum temperature is 140 °F (60 °C at 1.7 bar).