Hayward® True Union Ball Check Valves with Socket and NPT(F) Threaded Connectors
Toimittaja: Hayward
MFLX01341-10EA 229 EUR
MFLX01341-10 MFLX01368-10 MFLX01341-00 MFLX01341-02 MFLX01341-04 MFLX01341-08
Hayward® True Union Ball Check Valves with Socket and NPT(F) Threaded Connectors
Venttiilit Tarkista venttiilit
Unique square-cut elastomer seat provides seal at very low back pressures.
  • Constructed of a PVC body and EPDM seal
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.