ASCO Two- and Three-Way Miniature Solenoid Valves
Toimittaja: Asco
MFLX98515-32EA 63 EUR
MFLX98515-32 MFLX98515-42 MFLX98515-30 MFLX98515-40 MFLX98515-28 MFLX98515-38 MFLX98515-26 MFLX98515-36 MFLX98515-24 MFLX98515-34 MFLX98515-44 MFLX98515-22
ASCO Two- and Three-Way Miniature Solenoid Valves
Venttiilit Solenoidi venttiilit
Corrosion-resistant materials of construction.
  • Low power consumption
  • Manifold mount construction allows easy assembly
  • Ports are #10-32 UNF
Universal-operation, three-way valves allow you to introduce pressure at any port for gas or chemical mixing into the third port.