VWR Advanced magnetic stirrers are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With stirring speeds reaching 1600 rpm, VWR magnetic stirrers provide proper mixing. Available with ceramic plates.
- Excellent stirring
- Easy to read, large LCD displays speed
- Industry-leading safety features
- Cool touch and chemical resistant housing
- 5 year warranty
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays speed settings and intuitive icons.
Magnetic stirrers are available with ceramic top plate material. Reflective, white ceramic tops are chemical-resistant and easy to clean.
The low profile design utilises minimal bench space and allows use within a fume hood. Unit housing is constructed of a heat and chemical resistant polymer that remains cool to the touch during operation. The spill resistant design channels fluids away from internal components in the event of a spill.
Corresponding magnetic hotplate stirrers (heating and stirring 442-1265, 442-1268, 442-1270 and 442-1274) or hotplates (heating only 442-1269) with ceramic plates are also available.
Sertifikaatit: RoHS compliant. TUV listed.
Toimitustiedot: Units are supplied with a 234 cm three-wire cord and plug.