Vitality hrGFP II mammalian expression vectors
Toimittaja: Agilent Technologies
AGLS240144EA 845 EUR
AGLS240144
Vitality hrGFP II mammalian expression vectors
Vektorit, plasmidit ja kirjastot
Vitality hrGFP II Mammalian Expression Vectors support a variety of expression configurations, thus providing ideal expression options for each specific application.
  • Easy to detect
  • Applications include Western blotting, immunoprecipitation and flow cytometry
This is the next generation of original hrGFP that fluoresces brighter in mammalian cells is less toxic than EGFP, making it easier to use for mammalian expression experiments. Since hrGFP is derived from a different organism than EGFP, the hrGFP antibody does not cross react with the Aequorea Victoria GFP variant-derived EGFP.
Varoitus: For research use only. Not for use in diagnostic procedures.