Abiraterone Acetate

Toimittaja: Thermo Scientific
Varoitus

ACRO466730010EA 299 EUR
ACRO466730010
Abiraterone Acetate
Abiraterone Acetate
Kaava: C₂₆H₃₃NO₂
MW: 391,56 g/mol
Sulamispiste: 147°C
Storage Temperature: Pakastin
MDL numero: MFCD00934213
CAS-numero: 154229-18-2

Order Now

Testitulosten erittely

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Melting point 143.0°C to 147.0°C
HPLC ≥ 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR