VWR®, EPA 537.1 Method Standard, CRM
Toimittaja: Avantor
VWR®
89084.010EA 499 EUR
89084.010
VWR®, EPA 537.1 Method Standard, CRM
Standardiaineet ja vertailumateriaalit Ympäristöanalytiikan standardit ja vertailumateriaalit
This updated version of Method 537 can be used for the quantitative analysis of 18 analytes by Solid Phase Extraction (SPE) and Liquid Chromatography/Tandem Mass Spectrometry (LC/MS/MS).
Contains:
Perfluoro(2-methyl-3-oxahexanoic) acid (CAS 13252-13-6)
N-ethylperfluoro-1-octanesulfonamidoacetic acid (linear) (CAS 2991-50-6)
N-methylperfluoro-1-octanesulfonamidoacetic acid (linear) (CAS 2355-31-9)
Perfluorobutane-1-sulfonic acid (CAS 375-73-5)
Perfluoro-n-decanoic acid (CAS 335-76-2)
Perfluoro-n-dodecanoic acid (CAS 307-55-1)
Perfluoro-n-heptanoic acid (CAS 375-85-9)
Perfluorohexane-1-sulfonic acid (Linear and Branched) (CAS 355-46-4)
Perfluoro-n-hexanoic acid (CAS 307-24-4)
Perfluoro-n-nonanoic acid (CAS 375-95-1)
Perfluorooctane-1-sulfonic acid (Linear and branched) (CAS 1763-23-1)
Perfluoro-n-octanoic acid (CAS 335-67-1)
Perfluoro-n-tetradecanoic acid (CAS 376-06-7)
Perfluoro-n-tridecanoic acid (CAS 72629-94-8)
Perfluoro-n-undecanoic acid (CAS 2058-94-8)
11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid (CAS 763051-92-9)
9-Chlorohexadecafluoro-3-oxanone-1-sulfonic acid (CAS 756426-58-1)
4,8-Dioxa-3H-perfluorononanoic acid (CAS 919005-14-4)
Sertifikaatit: ISO 17034