WHEATON® DUALFUSION vial combine the best properties of glass, with the mechanical strength and the precision molding of plastic.
- Stand-alone nested tubs can be paired with snap cap and aluminum seal closure offerings
These vial combine the best properties of glass, with the mechanical strength and the precision molding of plastic. The vial are engineered to protect pharmaceutical drugs and sample formulations that are prone to cause glass delamination and that are sensitive to oxidation via the permeation of atmospheric oxygen. Using a state-of-the-art Plasma Enhanced Chemical Vapor Deposition (PECVD) Technology, an organosilicate protective layer is fused with a silica-like (SiO₂ barrier) layer. These layers are fused to a Cyclic Olefin Polymer (COP) shell to form a robust covalently bonded material. The result is an inert surface that offers chemical resistance and unparalleled protection against container leachates and environmental stressors, such as oxygen.
These vial are manufactured by and sold under license from SiO₂ Medical Products, Inc.
Sertifikaatit: Manufactured in a Class 7 Clean Room and E-Beam sterilized to meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injectables.
Toimitustiedot: DUALFUSION vial available in stand-alone nested tubs or as part of a COMPLETEPAK offering.
COMPLETEPAK configurations include closures and CoA.