Purified chemically synthesised sgRNA, CRISPR SureGuide
Toimittaja: Agilent Technologies
AGLSG7250EEA 995 EUR
AGLSG7250E AGLSG7250D AGLSG7250C AGLSG7250B
Purified chemically synthesised sgRNA, CRISPR SureGuide
Agilent SureGuide Purified Chemically Synthesised CRISPR sgRNAs are the industry standard for research-grade RNA oligonucleotides.
  • Fully custom de novo synthesis for every guide, including the tracrRNA sequence
  • Proprietary chemistry and purification method for high quality and high reproducibility across sgRNA sequences and orders
  • Manufactured under ISO 13485
Agilent SureGuide Purified Chemically Synthesised CRISPR Single Guide RNAs (sgRNAs) are produced using fully custom de novo synthesis, including the target and tracrRNA sequences. SureGuide Purified sgRNAs are manufactured under ISO 13485 standards using a proprietary chemistry and purification method for a high quality, highly reproducible, and streamlined process.
SureGuide Purified sgRNAs are available in 90 to 120 nucleotide (nt) lengths and with up to 10 chemical modifications. Modifications include: 2’-O-methyl (M); 2’-O-methyl 3’-phosphorothioate (MS); Deoxy (D); or 2’-ribo 3’-phosphorothioate (S).
Tilaustiedot: Each sgRNA is user defined and custom. Please contact your represenative for a quote.
SureGuide purified sgRNAs
I want to be contacted by Life Science Specialist to get a quotation
QUOTE REQUEST