HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Powder
Toimittaja: HyClone products (Cytiva)
HYCLSH30014.04EA 0 EUR
HYCLSH30014.04 HYCLSH30014.02 HYCLSH30014.05
HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Powder
Biokemialliset puskuriliuokset Puskurit soluviljelyyn
Balanced salt solution for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process.
  • Available in lot sizes that facilitate scale-up and manufacturing.
  • Without Sodium bicarbonate
  • Non-sterile
Sertifikaatit: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Varoitus: Storage 2° to 8° C