2-Amino-5-methyl-4-phenylthiazole 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD00746956-1GEA 65.5 EUR
BLDPBD00746956-1G BLDPBD00746956-5G BLDPBD00746956-250
2-Amino-5-methyl-4-phenylthiazole 98%
2-Amino-5-methyl-4-phenylthiazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₀H₁₀N₂S
MW: 190,2648 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00617257
CAS-numero: 30709-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR