3-(Benzyloxy)-5-chloro-1-ethylpyridin-2-one 96%
|
|
BLDPBD00752675-100EA
84.2
EUR
BLDPBD00752675-100
BLDPBD00752675-250
BLDPBD00752675-1G
BLDPBD00752675-5G
3-(Benzyloxy)-5-chloro-1-ethylpyridin-2-one 96%
3-(Benzyloxy)-5-chloro-1-ethylpyridin-2-one
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₄H₁₄ClNO₂ MW: 263,7200 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD30834429 CAS-numero: 2140305-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...