Imidazo[1,2-a]pyridin-8-ylmethanamine 95%

BLDPBD00754524-100EA 188 EUR
BLDPBD00754524-100 BLDPBD00754524-50M BLDPBD00754524-250
Imidazo[1,2-a]pyridin-8-ylmethanamine 95%
Imidazo[1,2-a]pyridin-8-ylmethanamine
Kaava: C₈H₉N₃
MW: 147,18 g/mol
MDL numero: MFCD10698077
CAS-numero: 933721-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR