2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine 98% (by GC, titration analysis)

BLDPBD00786056-100EA 23.9 EUR
BLDPBD00786056-100 BLDPBD00786056-250 BLDPBD00786056-1G
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine 98% (by GC, titration analysis)
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₅F₃N₄
MW: 214,15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD26403657
CAS-numero: 219508-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR