Sodium-2,4-dimethylbenzenesulphonate monohydrate 98% (by titrimetric analysis)

BLDPBD00803976-100EA 108 EUR
BLDPBD00803976-100 BLDPBD00803976-5G BLDPBD00803976-25G
Sodium-2,4-dimethylbenzenesulphonate monohydrate 98% (by titrimetric analysis)
Sodium-2,4-dimethylbenzenesulphonate monohydrate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₈H₁₀O₃S
MW: 208,21 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD15071080
CAS-numero: 142063-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR