Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate 98%

BLDPBD00811202-25GEA 647 EUR
BLDPBD00811202-25G BLDPBD00811202-5G BLDPBD00811202-1G
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate 98%
Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₉H₂₂F₃IO₃S
MW: 514,34 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD25541454
CAS-numero: 139139-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR