(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid 98%

BLDPBD00833633-1GEA 502 EUR
BLDPBD00833633-1G BLDPBD00833633-250 BLDPBD00833633-100
(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid 98%
(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₆F₁₇O₃P
MW: 528,10 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD19105612
CAS-numero: 80220-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR