2-Biphenylboronic acid pinacol ester 98%
|
|
BLDPBD00903472-1GEA
33.3
EUR
BLDPBD00903472-1G
BLDPBD00903472-25G
BLDPBD00903472-5G
2-Biphenylboronic acid pinacol ester 98%
2-Biphenylboronic acid pinacol ester
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₁₈H₂₁BO₂ MW: 280,17 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD14636459 CAS-numero: 914675-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...