5,10,15,20-Tetrakis(1-methylpyridinium-4-yl)porphyrin tetrachloride 95%

BLDPBD00934015-250EA 231 EUR
BLDPBD00934015-250 BLDPBD00934015-1G BLDPBD00934015-100
5,10,15,20-Tetrakis(1-methylpyridinium-4-yl)porphyrin tetrachloride 95%
5,10,15,20-Tetrakis(1-methylpyridinium-4-yl)porphyrin tetrachloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₄₄H₃₈Cl₄N₈
MW: 820,6400 g/mol
Storage Temperature: Huoneenlämpö
CAS-numero: 92739-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR