3-(2-Pyridylcarbamoyl)phenylboronic acid 95%
|
|
BLDPBD00936227-1GEA
305
EUR
BLDPBD00936227-1G
BLDPBD00936227-250
BLDPBD00936227-100
3-(2-Pyridylcarbamoyl)phenylboronic acid 95%
3-(2-Pyridylcarbamoyl)phenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₁₂H₁₁BN₂O₃ MW: 242,0383 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD28167454 CAS-numero: 874459-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...