1-Bromo-3-(trimethylsilyl)benzene 95%

BLDPBD00953893-1GEA 21.9 EUR
BLDPBD00953893-1G BLDPBD00953893-25G BLDPBD00953893-5G
1-Bromo-3-(trimethylsilyl)benzene 95%
1-Bromo-3-(trimethylsilyl)benzene

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₉H₁₃BrSi
MW: 229,1890 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00093882
CAS-numero: 17878-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR