Tetrabutylammonium hydrogencarbonate 95%

BLDPBD00973756-1GEA 67.6 EUR
BLDPBD00973756-1G BLDPBD00973756-5G BLDPBD00973756-250
Tetrabutylammonium hydrogencarbonate 95%
Tetrabutylammonium hydrogencarbonate
Kaava: C₁₇H₃₇NO₃
MW: 303,48 g/mol
MDL numero: MFCD20134057
CAS-numero: 17351-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR