2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD01084546-25GEA 1160 EUR
BLDPBD01084546-25G BLDPBD01084546-5G BLDPBD01084546-1G BLDPBD01084546-250
2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%
2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₁₁H₁₁BINO₄
MW: 358,925 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD11215234
CAS-numero: 1287221-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR