5-(Difluoromethyl)-3-methyl-1H-pyrazole 98%
|
|
BLDPBD01109239-5GEA
398
EUR
BLDPBD01109239-5G
BLDPBD01109239-1G
BLDPBD01109239-250
5-(Difluoromethyl)-3-methyl-1H-pyrazole 98%
5-(Difluoromethyl)-3-methyl-1H-pyrazole
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₅H₆F₂N₂ MW: 132,11 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD08701204 CAS-numero: 934759-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...