2-Methoxycinnamic acid 98% + (predominantly trans isomers)

BLDPBD01391964-25GEA 47.8 EUR
BLDPBD01391964-25G BLDPBD01391964-500 BLDPBD01391964-100 BLDPBD01391964-0.0 BLDPBD01391964-5G
2-Methoxycinnamic acid 98% + (predominantly trans isomers)
2-Methoxycinnamic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₀O₃
MW: 178,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064238
CAS-numero: 6099-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR